ContactUsLinkComponent
XRF1920-760.jpg
X-Ray Fluorescence Spectroscopy (XRF)
XRF | Avantor
X-ray fluorescence spectroscopy, or XRF, lets analysts obtain qualitative and quantitative information by measuring the fluorescent X-rays that are emitted from a sample when X-rays are projected onto the sample from an outside source. Avantor® can provide you with the products and materials you need to conduct XRF in your lab. Choose the products that work best for you from our extensive selection.
Spectromelt® fluxes for fusion sample preparation. Lithium borates, Spectromelt A, sodium borates and Spectromelt B10 are available.
Literature