ContactUsLinkComponent
mass_spectrometry_hero.jpg
Mass Spectrometry
Mass spectrometry is an important tool when specific compounds and materials must be identified with a high degree of precision. Among its uses are food quality and safety, drug testing, analyzing pesticide residue, protein identification via peptide mapping and carbon dating. Avantor® offers a comprehensive range of products to meet your mass spectrometry needs. 
Mass Spectrometry | Avantor
Questions about our mass spectrometry products?
Contact us today for expert technical advice and to discuss your LC and GC needs. Our team is ready to help at [email protected]
Contact us for more information and free technical advice
Featured Products
GC_MS_Columns.jpg
For your GC-MS applications, low bleed stationary phases are very important. Avantor provides a wide range of polarity GC phases designed for mass spec applications.  
LC_MS_Grade_Solvents_new.jpg
For your LC-MS applications, solvents that feature high UV transmittance, low acidity and alkalinity, and low particulates ensure minimal interference/background in MS traces.
Mass Spectrometry Products
MassSpectrometry500.jpg
Avantor offers a comprehensive line of protein reagents to use with protein assays to measure the total protein concentration of a sample compared to a standard. See what we have to offer. 
Chromatography_500.jpg
Chromatography columns are an essential tool for separating compounds from a sample mixture to allow for specific sample collection. Avantor carries different selectivity columns so you can find the one that’s best for your application. 
ProteinsAndPeptides500.jpg
Avantor can provide your lab with a variety of kits, mixture solutions and collagen matrices that you can use for many essential laboratory functions. 
OtherProducts500.jpg
Avantor carries a wide range of equipment, products and supplies commonly used in mass spectrometry. Take a look at everything we have to offer. 
Rapid High throughput LC-MS/MS separations are achievable using a new format column, find out more here.
Mass Spectrometry Explained
Mass spectrometry is an important tool for identifying specific compounds or materials with a high degree of precision. This technique has many applications ranging from food quality and safety to carbon dating.
In the most basic terms, mass spectrometry is a sensitive technique used to detect, identify and quantitate molecules based on the molecule’s mass-to-charge (m/z) ratio.
How it Works
All mass spectrometers have three primary components; an ion source, a mass analyzer, and an ion detector. Samples are prepared and loaded into the mass spectrometer in either liquid, gas, or dried form and are then vaporized and ionized by the ion source. There are variations of these components in each type of mass spectrometer, which offer a diverse range of options for testing different physical properties of samples and data collection.
Mass Spectrometry Applied
There are numerous applications for mass spectrometry, including both qualitative and quantitative uses. Mass spectrometry is most commonly used in analytical laboratories for the study of physical, chemical, or biological properties of compounds.
Additional applications of mass spectrometry:
  • Identifying unknown compounds
  • Determining the isotopic composition of elements in a molecule
  • Determining the structure of a compound based on fragmentation
  • Quantifying the amount of a compound in a sample
  • Understanding the fundamentals of gas phase ion chemistry
This makes mass spectrometry an ideal tool in drug discovery, clinical testing, genomics, geology and environmental studies on food and soil. Among the many applications in medicine and genomics, mass spectrometry can be used for the characterization of single nucleotide polymorphisms (SNPs), as well as in rapid toxic screenings for detecting toxins in blood.
Are you ready to move your next innovation forward? We can help.
Contact us to speak with a member of our team.
Contact us for more information and free technical advice