ContactUsLinkComponent
Liquid-Chromatography-Analytical1920-760.jpg
Liquid Chromatography: Analytical
Liquid Chromatography - Analytical | Avantor
High-performance liquid chromatography (HPLC) and ultra-high-performance liquid chromatography (UHPLC) are among the most widely used techniques in the chemical and life sciences laboratories. Both are important techniques that have many scientific, academic and industrial applications. Avantor® has a comprehensive range of equipment, consumables, products and services to support analytical liquid chromatography assays in your laboratory.
Questions about our analytical liquid chromatography products?
Contact us today to discuss your needs. Our team is ready to help.
Contact us for more information and free technical advice
Analytical Liquid Chromatography Products
Column-Selectiom400-495..jpg
You need the right HPLC or UHPLC column for your analytes. There are many options out there. We can save you time by helping you to select the right one for your application.
Stationary-Phases400-495.jpg
Avantor carries thousands of chromatography columns, you can find the products that best meet your needs using our easy column selection tool. Search our database to find columns that match your specific method requirements.
Mobile-Phases-Modifiers400-495.jpg
You can find the HPLC solvents and modifiers that are most suitable for your assays from the hundreds of products Avantor offers. With different grades available to meet your needs, we have a solution for all types of analytical method.
Vials400-495.jpg
No matter what type of vials you need, Avantor can help. We have dozens of vials of different sizes, shapes and finishes to help guarantee chemical compatibility. Look at what Avantor carries to meet your needs.
Expert-Chromatography-260x190.jpg
Avantor® - Experts in Chromatography
Do you need technical information or advice? We have the technical information and experts to help.
Contact us
What is HPLC?
High performance liquid chromatography, known as HPLC, works by using high-pressure pumps to effectively force a sample mixture containing the solvent through a column that contains a solid absorbent material.
This absorbent material (commonly silica) interacts differently with each component in the solvent. As the solvent components flow out of the column, they migrate through the column at a unique rate.
The difference in migration results in each component being separated from the mixture. A detector then generates a peak for each component and can allow us to determine the identity and concentration of the compounds.
Are you ready to move your next innovation forward? We can help.
Contact us to speak with a member of our team.
Contact us for more information and free technical advice