Analytik Jena PCR and Real-Time PCR Thermal Cyclers
Analytik Jena offers a wide product variety of high-performance standard thermal cyclers for PCR as well as highly efficient real-time thermal cyclers for DNA amplification and quantification.
qTOWER3 Real-Time PCR
The top performer among real-time PCR thermal cyclers, capable of scanning an entire plate in only six seconds.
qTOWER3 84 High-Throughput Real-Time PCR
High-performance systems for high-throughput applications using real-time PCR in a 384-sample multiwell format.
Biometra PCR Thermal Cyclers
The Biometra product family ranges from standard to premium equipment, designed with the common principles of being reliable, convenient, and precise.