Masterflex® L/S & I/P Drives with Advanced Networking & Communication
More Pump Control Than Ever
New features for select Masterflex L/S & I/P drives
- Ethernet/IP options
- Profibus options
- Bluetooth
- Cloud-based monitoring
All units are equipped with Masterflex open-head sensor technology, are able to accept the full range of Masterflex® L/S® or I/P® pump heads, and use tubing from the broadest selection of performance materials in the industry.
Process Automation Controls
Effortlessly integrate your pump into existing systems
- Common Industrial Protocol via Ethernet/IP
- Compatible with today’s advanced automated control systems
- Electronic Data Sheet (EDS) provided at no extra cost
Remote monitoring
Monitor your pump remotely
- Monitor your pump from an iOS-enabled tablet or smartphone via Bluetooth
- Cloud monitoring through Ethernet connection for pump verification and alarms
- USB protocols for PC connectivity and communications
Need more information or help selecting the perfect product for your application?
Our experts are ready to help. Contact us at 1-800-MASTERFLEX or sales@masterflex.com