Masterflex® Peristaltic Pump Tubing
Accept no substitutes —
Precision extruded & pump tested formulations engineered to provide the highest quality for tubing life, pressure, and suction lift.
Masterflex brand pump tubing is custom extruded to fit Masterflex pumps and engineered for long life in peristaltic pump applications. Lot-to-lot consistency provides accuracy and repeatability.
Shop by pump tubing series
View Masterflex Pump Tubing Selection Guide
Top Tubing Formulations
Silicone
The biopharmaceutical industry standard
C-Flex
For aseptic sealing, welding, and other biologic applications
Puri-Flex
Superior pump life and biocompatibility. Stock guaranteed
PharMed BPT
Trusted for peristaltic and pharmaceutical applications
PTFE
Absolute chemical resistance and high heat resistance
Chem-Durance
Excellent chemical resistance, pump life, and biocompatibility
Tygon E-LFL
Longest pump life of all Tygon formulations
Viton
Ideal for required FDA compliance and superior chemical compatibility.
Custom services and tools
Custom-engineered solutions
Partnering on custom engineered pumps – making ideas reality for over 50 years.
Learn more
Custom cut-to-length tubing
We have the flexibility to meet your needs and save you time and money. Choose your tubing type, length and quantity.
Learn more
Other featured categories
How to verify your tubing compatibility
Weigh and Measure
the length and diameter of tubing sample
Immerse
Using a closed vessel, immerse the sample in your fluid for 48 hours
Reweigh, Remeasure,
and examine the sample for swelling, brittleness or other deterioration
Test in Pump
under actual temperature, vacuum/pressure, or other operating conditions
NOTE: PharmaPure and Chem-Durance Bio tubing formulations are co-extruded with different inner and outer materials and require modified testing methods. Contact Masterflex®, part of Avantor for chemical testing instructions. Check our Chemical Resistance Database.
Need more information or help selecting the perfect product for your application?
Our experts are ready to help. Contact us at 1-800-MASTERFLEX or sales@masterflex.com