Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Gram Stain Sets and Reagents

Supplier: VWR International

Gram Stain Sets include all the necessary solutions to fully differentiate Gram positive and Gram negative specimens.

  • Sets include all necessary solutions
  • Enables differentiation
All Gram stain and replacement sets are packaged in a styrofoam tray and come with spout caps for ease of pouring.



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All

Reagents
Description Size VWR Catalog Number Unit Price Quantity
Crystal Violet 3.8 L (1 gal.) 10143-260 Case of 4 (1 US Gal) Retrieving Restricted
Each (1US Gal) Retrieving Restricted
1:1 Decolorizer 3.8 L (1 gal.) 10143-602 Case of 4 (1 US Gal) Retrieving Restricted
Each (1US Gal) Retrieving Restricted
Decolorizer 3.8 L (1 gal.) 10143-264 Case of 4 (1 US Gal) Retrieving Restricted
Each (1US Gal) Retrieving Restricted
Iodine Lugol's Working 3.8 L (1 gal.) 10143-262 Case of 4 (1 US Gal) Retrieving Restricted
Each (1US Gal) Retrieving Restricted
Safranin 3.8 L (1 gal.) 10143-604 Case of 4 (1 US Gal) Retrieving Restricted
Each (1US Gal) Retrieving Restricted
Stabilized Iodine 3.8 L (1 gal.) 10143-180 Case of 4 (1 US Gal) Retrieving Restricted
Each (1US Gal) Retrieving Restricted

Replacement Sets
Description Size VWR Catalog Number Unit Price Quantity
Crystal Violet Replacement Set 4 x 237 mL (8 oz.) 10143-168 Case of 6 (EA) Retrieving Restricted
Kit Retrieving Restricted
Crystal Violet Replacement Set 4 x 474 mL (16 oz.) 10143-182 Case of 4 (EA) Retrieving Restricted
Kit Retrieving Restricted
1:1 Decolorizer Replacement Set 4 x 237 mL (8 oz.) 10143-174 Case of 6 (EA) Retrieving Restricted
Kit Retrieving Restricted
1:1 Decolorizer Replacement Set 4 x 474 mL (16 oz.) 10143-252 Case of 4 (EA) Retrieving Restricted
Kit Retrieving Restricted
Decolorizer Replacement Set 4 x 237 mL (8 oz.) 10143-172 Case of 6 (EA) Retrieving Restricted
Kit Retrieving Restricted
Decolorizer Replacement Set 4 x 474 mL (16 oz.) 10143-184 Case of 4 (EA) Retrieving Restricted
Kit Retrieving Restricted
Gram Stain Set 4 x 237 mL (8 oz.) 10143-178 Case of 6 (EA) Retrieving Restricted
Kit Retrieving Restricted
Gram Stain Set 4 x 474 mL (16 oz.) 10143-258 Case of 4 (EA) Retrieving Restricted
Kit Retrieving Restricted
Safranin Replacement Set 4 x 237 mL (8 oz.) 10143-176 Case of 6 (EA) Retrieving Restricted
Kit Retrieving Restricted
Safranin Replacement Set 4 x 474 mL (16 oz.) 10143-254 Case of 4 (EA) Retrieving Restricted
Kit Retrieving Restricted
Stabilized Iodine Replacement Set 4 x 237 mL (8 oz.) 10143-170 Case of 6 (EA) Retrieving Restricted
Kit Retrieving Restricted
Stabilized Iodine Replacement Set 4 x 474 mL (16 oz.) 10143-256 Case of 4 (EA) Retrieving Restricted
Kit Retrieving Restricted
restricted You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty