Would you like to log in again?
BDH HPLC Solvents
BDH HPLC Solvents
BDH HiPerSolv CHROMANORM high purity solvents for HPLC applications are manufactured at a state of the art European ISO 9001 - ISO 14001 solvent production facility. These solvents provide an unparalleled combination of performance and price for even the most demanding liquid chromatography applications. Packaged in standard 4L North American bottles and 4 x 4L cases, these solvents:
  • Are designed to meet demanding HPLC and UV/Vis analytical and quality control requirements
  • Are filtered down to 0.2 micron level and bottled under Nitrogen
  • Include super-gradient grade Acetonitrile, with >95% UV transmittance at 200 nm and <30 ppm water content
  • Feature tamper-evident caps for added peace of mind
  • Carry Globally Harmonized System (GHS) labeling to help with compliance
  • Feature environmentally-friendly corrugate/pulp case inserts which are fully recyclable
Browse our complete line of BDH Chemicals.
Shop our special offers:
Browse Products