Would you like to log in again?
Solvents, HPLC Grade, BDH
Solvents, HPLC Grade, BDH
BDH® Chemicals - Here We G-R-O-W Again!
Introducing BDH HiPerSolv CHROMANORM Solvents for HPLC
VWR International is proud to introduce the newest addition to the exclusive family of high-quality, competitively-priced BDH Chemicals - high purity solvents for HPLC applications. Manufactured at a state of the art European ISO 9001 - ISO 14001 solvent production facility, the initial range of six of the most popular HPLC solvents provides an unparalleled combination of performance and price for even the most demanding liquid chromatography applications. Packaged in standard 4L North American bottles and 4 x 4L cases, these solvents:
  • Are designed to meet demanding HPLC and UV/Vis analytical and quality control requirements
  • Are filtered down to 0.2 µm level and bottled under Nitrogen
  • Include super-gradient grade Acetonitrile, with >95% UV transmittance at 200 nanometers and <30ppm water content
  • Feature tamper-evident caps for added peace of mind
  • Carry Globally Harmonized System (GHS) labeling to help with compliance
  • Feature environmentally-friendly corrugate/pulp case inserts which are fully recyclable
Product Selection