Would you like to log in again?
Compounding Pharmacies
Compounding Pharmacies VWR® has the answers you need…
VWR offers a combination of product choice, differentiated services and world class logistics to bring you a complete solution. Lean on VWR for your compounding supply needs.
VWR® has your total solution for Compounding Pharmacies. Our team of Critical Environment Specialists are ready to provide expert advice as well as consultative services to help your organization comply with USP 797 and USP 800 guidelines. We understand your need to maintain the sterility of your environment to ensure regulatory compliance and ultimately patient safety.
Product Solutions
Featured Supplier
Our extensive product line for controlled environments includes all types of wipes, presaturated wipes, sterile and non-sterile wipes, mopping systems, wall washing systems and disinfectants.
Learn More
Featured Product
SANIHOL® ST Sterile 70% EtOH Solution
Sterile, ready-to-use 70% denatured ethanol (EtOH) solution is ideal for cleanroom use. Solution is filtered to 0.2µm to remove particulate contamination. It evaporates completely, leaving no residue.
Made according to USP specifications and packaged in an ISO Class 5 (FED-STD-209E Class 100/M3.5) cleanroom.
Learn More