Premier™ Pass-Thru Laboratory and Pharmacy Refrigerators, Nor-Lake® Scientific
Supplier: Horizon Scientific
Premier™
NSPT806WWW/0 NSPT524WWF/0 NSPT242WWG/0 NSPT806WWW/5 NSPT806WWG/0 NSPT524WWW/0 NSPT242WWF/0 NSPT524WWG/0 NSPT242WWW/0 NSPT806WWF/0
89368-288EA 42488.73 CAD
89368-288 89368-296 89368-190 89368-290 89368-198 89368-284 89368-292 89368-194 89368-280 89368-300
Premier™ Pass-Thru Laboratory and Pharmacy Refrigerators, Nor-Lake® Scientific
Refrigerators
Refrigerators are equipped with a digital LED display and microprocessor temperature controller with audible and visual Hi/Lo temperature alarm
All 115V models are supplied with a 5-15P NEMA plug and 15-15R NEMA receptacle. The 230V models feature a Power Inlet (IEC60320) Module and require a locally supplied cord.
Units offer adjustable temperature control from 2 to 10°C (35.6 to 50°F). They are factory preset at 4°C (39F). Safety features include, remote alarm contacts, continuous product temperature display, product sensor with glycerin bottle, and power supply switch. The exterior cabinet front, sides, and back are painted white. Exterior cabinet top and bottom are galvanized steel. Interior cabinet liner and door panels are painted white. Cabinet and solid doors are foamed-in-place with CFC-free, high-density polyurethane foam insulation. Glass door(s) on applicable models are full-size, double-pane with frame anti condensate heater wires. Glass door(s) are standard on all WWF suffix models and WWG suffix models (controller side). Solid door(s) on applicable models are full-size. Solid door(s) are standard on WWW suffix models and WWG suffix models (non-controller side). The heavy-duty doors feature pivot hinges, pull-door handles, magnetic door gaskets, and centered key door lock(s) (2 keys included). Additional standard features include, four casters (2 locking) on two- and four-door models, six casters (3 locking) on six-door models, three coated wire shelves per door section, interior light (switch activated), air cooled condensing unit, top-mounted forced air refrigeration system, automatic defrost, and condensate evaporation.
Ordering information: Refrigerators are supplied with an 18-month parts and labor warranty, 5-year compressor warranty (US and Canada), and 18-month parts warranty (International).