Variable Bore Tubing Connector, Ace Glass Incorporated
Supplier: Ace Glass
12714-20
89077-438PK 227.79 CAD
89077-438
Variable Bore Tubing Connector, Ace Glass Incorporated
Connectors Connectors, Tubing
This tubing connector enables the instant connection of rigid or flexible tubing within a range of 0.5 to 4mm O.D
Dead volume is zero. Connections are leak-tight to at least 50psi, and can be made in seconds without the flanging of tubing. Connector body is virgin PTFE. Caps are polypropylene with Fluoron elastomer O-Ring seals, and they feature ¼"–28 UNF thread for interfacing with adapters or other miniature plumbing systems. Supplied with PTFE cones for making O-Ring-free connections.