CoolRack® Sleeves, BioCision®
BCS-205
89369-134EA 43.97 CAD
89369-134
CoolRack® Sleeves, BioCision®
Colorful elastic sleeves brighten the lab and allow users to color-code experiments. Sleeves provide a comfortable grip when handling cold , warm, or wet CoolRack® tube modules. Sleeves are compatible with ice, dry ice, and water baths.
  • Sleeves are designed to fit all 15-Well CoolRack® modules, all 30-Well CoolRack® modules, CoolRack® 15ml, CoolRack® 50ml, CoolRack® VS13, CoolRack® V13, and CoolRack® V16.
Ordering information: Sleeves are supplied in packs of four, with one color each of purple, green, orange, and pink.