Adhesive Temperature Strips, BioCision®
BCS-143
89205-940PK 81.31 CAD
89205-940
Adhesive Temperature Strips, BioCision®
Liquid-crystal temperature display strips measure the temperature of a surface with 1°C resolution
Ideal for placement on CoolRack®, CoolSink™, and ThermalTray™ thermo-conductive modules. Strips feature permanent adhesive backing. They are not removable or autoclavable.
Ordering information: For CoolRack® thermo-conductive modules, see 95045-464 series. For CoolSink™ thermoconductive plate modules, see 95045-472 series. For ThermalTray™ thermo-conductive platforms, see 95045-448 series.