HPLC Column Connector, Single End Unit with .007" ID Tubing, Black, MicroSolv
49807-30 49807-06 49807-20 49807-10-S 49807-50-S 49807-20-S 49807-10 49807-06-S 49807-30-S
97056-694EA 243.38 CAD
97056-694 97056-692 97056-708 97056-706 97056-690 97056-704 97056-700 97056-698 97056-696
HPLC Column Connector, Single End Unit with .007" ID Tubing, Black, MicroSolv
These devices are permanently installed on your HPLC system and they allow you to use any HPLC Column with different fittings on that system without changing your end fittings
This makes your system fully compatible with columns with Waters®, Valco®, Parker® or other end fittings; no male nut changes are needed. The spring loaded mechanism self adjusts for proper fit and elimination of dead volume. Once you try them, you will see why high output labs use these routinely. Connects from outlet to column and from the column to the detector.
Ordering information: Each connector is supplied with color-coded bands to identify the ID of the tubing. Available with PEEK Tubing. Other sizes available.