Select™ Sliding Glass Door Laboratory and Pharmacy Refrigerators, Nor-Lake® Scientific
Supplier: Horizon Scientific
NSSR502WWG/0
89368-362EA 20826.86 CAD
89368-362
Select™ Sliding Glass Door Laboratory and Pharmacy Refrigerators, Nor-Lake® Scientific
Refrigerators are equipped with a digital LED display microprocessor temperature controller, with audible and visual Hi/Lo temperature alarm
All 115V units are supplied with a 5-15P NEMA plug and 5-15R NEMA receptacle. The 208/230V units are supplied with a 6-15P NEMA plug and 6-15R NEMA receptacle. The 230V units feature a Power Inlet module (IEC60320) and require a locally supplied cord.
Temperature control is adjustable from 2 to 10°C (35 to 50°F). Units are factory preset at 4°C (39°F). Safety features include, remote alarm contacts, continuous product temperature display, product sensor with glycerin bottle, and power supply switch. The exterior cabinet front, sides, and back are painted white. Exterior cabinet top and bottom are galvanized steel. Interior cabinet liner is painted white. Cabinet is foamed-in-place with CFC-free, high-density polyurethane foam insulation. Glass doors are full size and double pane. Self-closing glass doors have a hold-open feature and centered key door lock(s) (2 keys included). Units also include, four casters (2 locking), three coated wire shelves per door section, interior light (switch activated), air cooled condensing unit, and top-mounted forced air refrigeration system.
Certifications: UL and C-UL listed.
Ordering information: Refrigerators are supplied with 24-month parts and labor warranty, 5-year compressor warranty (US and Canada), and 18-month parts warranty (International).