VWR® Universal Specific Gravity and Baume Plain Form Hydrometer, Traceable to NIST

Supplier: VWR, part of Avantor
61806-0011
34675-008EA 110.52 CAD
34675-008
VWR® Universal Specific Gravity and Baume Plain Form Hydrometer, Traceable to NIST
Hydrometers
Used in the chemistry, pharmacology, food and beverage, and starch industries to name a few, these dual scale, plain form hydrometers measure specific gravity/relative density (g/cm3) of liquids lighter and heavier than water and Baume.

  • Lead free ballast and glass
  • Individually serialized
  • Temperature standardization: of 60 °F (15 °C)
  • Hand inspected by our U.S. quality team

Certifications: Traceable to NIST.

Packaging: Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy.

Caution: Hydrometers should be stored in a clean, dry, safe place, and should not be subjected to extreme temperatures.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR