Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "line-valves"

https://ca.vwr.com/stibo/smallweb/std.lang.all/94/80/18989480.jpg

Line valves, or single-stage regulators, are designed to limit the flow of liquids or gases to one direction. Used for instrumentation, hydraulic pipes, pneumatic pipes, chromatography, control panels, moderate vacuum systems, chemical research, gas analysis, and more. Excellent diaphragm sealing ensures tightness against leaking. Our selection of line valves requires no special tools to assemble, and they are easy to assemble, fit, and tighten.

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"9"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  Ace Glass
Description:   Used as a one-time opening valve. Drawn to a point so that a small weight will fracture seal.
Product available on GSA Advantage® Small Business Enterprise
Supplier:  Ace Glass
Description:   This flask is supplied with a 1" beaded pipe-type flange extension off bottom for connection to a 0–20 mm threaded PTFE stopcock as the outlet. Stopco...
Product available on GSA Advantage® Small Business Enterprise
Supplier:  Ace Glass
Description:   Redesigned flush-seal type valve with the top of the valve having a 1-1/2" x 3/8" O.D. pin that extends upward. The pin fits into the receptacle on th...
Product available on GSA Advantage® Small Business Enterprise
Supplier:  Ace Glass
Description:   A water or air shut-off solenoid valve for use with water-flo power cut-off or as a general laboratory shut-off valve. Operates from 0.35Kg/cm2
Product available on GSA Advantage® Small Business Enterprise
Catalog Number: (89130-356)
Supplier:  Ceodeux Inc.
Description:   Used for instrumentation, hydraulic pipes, pneumatic pipes, chromatography, control panels, moderate, vacuum systems, chemical research, gas analysis,...
Supplier:  Ceodeux Inc.
Description:   Low pressure line valve for various pure gases. High leak tightness through diaphragm sealing.
Supplier:  Ceodeux Inc.
Description:   Low pressure line valve for various pure gases. High leak tightness through diaphragm sealing.
Supplier:  Restek
Description:   Static and pressure relief valves are available for use with multiple ASE® systems.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 9
  1  2  Next