Anti-5HT2C Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX77943
89366-436EA 699.51 CAD
89366-436
Anti-5HT2C Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to 5-HT2C Receptor

Type: Primary
Antigen: 5HT2C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR