Anti-5HT7R Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX78121
89366-510EA 701.31 CAD
89366-510
Anti-5HT7R Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to 5-HT7 Receptor

For IF: Use at a concentration of 13 ?g/ml. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: 5HT7R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR