Anti-5HT7R Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX70937
89365-144EA 699.51 CAD
89365-144
Anti-5HT7R Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal to Human 5 - HT7 Receptor

Immunohistochemistry with formalin fixed paraffin embedded tissues. Not tested for other uses. IHC-P: Use at a dilution of 1/100 - 1/200.

Type: Primary
Antigen: 5HT7R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR