Anti-Bovine RESPIRATORY SYNCYTIAL Virus Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX70322
89364-910EA 565.62 CAD
89364-910
Anti-Bovine RESPIRATORY SYNCYTIAL Virus Rabbit Polyclonal Antibody
Antibodies
Human Respiratory Syncytial Virus (RSV) and Bovine Respiratory Syncytial Virus (BRSV) Fusion protein of all strains of RSV and BRSV.

Type: Primary
Antigen: Bovine RESPIRATORY SYNCYTIAL Virus
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR