Anti-C3C Sheep Polyclonal Antibody

Supplier: GeneTex
GTX74127
89367-756EA 881.46 CAD
89367-756
Anti-C3C Sheep Polyclonal Antibody
Antibodies
Sheep polyclonal antibody to C3c

Recommended Starting Dilutions: Immunohistology - Frozen: 1/50 - 1/100 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: C3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR