Anti-CALCR Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX78139
89366-516EA 699.51 CAD
89366-516
Anti-CALCR Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Calcitonin Receptor

Type: Primary
Antigen: CALCR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR