Anti-IgG Goat Polyclonal Antibody

Supplier: GeneTex

GTX77535
89353-772EA 252.5 CAD
89353-772
Anti-IgG Goat Polyclonal Antibody
Antibodies
HRP-conjugated Goat anti-Human IgG (H+L) polyclonal antibody

Optimal conjugate working dilutions should be determined for your particular assay conditions.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR