Anti-AKT1 Rabbit Polyclonal Antibody

GTX101073
89348-802EA 472.59 CAD
89348-802
Anti-AKT1 Rabbit Polyclonal Antibody
Rabbit Polyclonal antibody to AKT1 (v-akt murine thymoma viral oncogene homolog 1)

Suggested starting dilutions are as follows: WB: 1:1000-1:10000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: AKT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR