Anti-A1AT Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX20922
89351-586EA 565.64 CAD
89351-586
Anti-A1AT Rabbit Polyclonal Antibody
Antibodies
Alpha 1 Antitrypsin is a plasma protein synthesised in the liver. It consists of a single polypeptide chain and has a molecular weight of 52 kDa. A serine protease inhibitor, its primary function is the protection of lung tissue from the action of neutrophil elastase. The average level of alpha 1 Antitrypsin in plasma is 1.3 g/l.

An incubation of 30 minutes at RT is recommended. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: A1AT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR