Anti-ATM Rabbit Polyclonal Antibody

GTX74214
89367-764EA 630.78 CAD
89367-764
Anti-ATM Rabbit Polyclonal Antibody
Recognizes the human ATM protein, a 350kD polypeptide that is the product of the ATM gene. The ATM gene is defective in Ataxia telangiectasia. The antiserum has been termed ATM.V. in relevant publications.

Type: Primary
Antigen: ATM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR