Anti-GAPDH Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX124503
89359-448EA 762.37 CAD
89359-448
Anti-GAPDH Rabbit Polyclonal Antibody
Antibodies
Suggested starting dilutions are as follows: WB: 1:5000-1:20000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: gapdh
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Zebra Fish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR