Anti-GFAP Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX108711
89355-672EA 891.75 CAD
89355-672
Anti-GFAP Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to GFAP (glial fibrillary acidic protein)

Suggested starting dilutions are as follows: WB: 1:5000-1:20000, IHC-Fr: 1:100-1:1000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: GFAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR