Anti-IgG Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX77191
89353-532EA 228.71 CAD
89353-532
Anti-IgG Rabbit Polyclonal Antibody
Antibodies
The antibodies are specific for the Fc fragment of IgG based on IEP, gel diffusion and ELISA. Rabbits were immunized with highly purified goat IgG Fc prepared by papain digestion. The antiserum was solid phase absorbed with Fab fragments and the anti Fc antibodies immunoaffinity purified off an Fc immunoabsorbant.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(c)
Host: Rabbit
Isotype: IgG
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR