Anti-VEGF Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX74091
89365-380EA 1000.71 CAD
89365-380
Anti-VEGF Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to VEGF

Recommended Starting Dilutions:ELISA: Max Dilution 0.5 ug/ml Functional Assays: 1.0 ug/ml - 2.0 ug/ml Immunofluorescence: Min Dilution 2.5 ug/ml Western Blotting: 0.1 ug/ml - 0.2 ug/ml Immunohistology - Paraffin: Min Dilution 2.5 ug Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: VEGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR