Anti-VEGF Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX29570
89362-828EA 778.2 CAD
89362-828
Anti-VEGF Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to VEGF

Suggested concentration: Elisa: 2-3 ?g/ml; Neutralization: 5-10 ?g/ml; WB: 0.5-1 ?g/ml. However, the optimal conditions should be determined individually.

Type: Primary
Antigen: VEGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR