Anti-TRPV1 Rabbit Polyclonal Antibody
GTX78182
89366-534EA
454.38
CAD
89366-534
Anti-TRPV1 Rabbit Polyclonal Antibody
Rabbit polyclonal to VR1 N-terminus antibody
Immunohistochemistry 1:1000; Immunocytochemistry 1:1000 Western Blotting 1:1000 Dilutions listed only as a recommendation. Optimal dilution should be determined by investigator.Immunohistochemistry: Antiserum was used on perfusion fixed tissue. Perfusion: 1) calcium-free Tyrode’s solution, 2) fixative, and 3) 10% sucrose in PBS as a cryo-protectant. Desired tissues were dissected and stored overnight in 10% sucrose in PBS. Slide-mounted tissue sections were processed for indirect immunofluorescence. Slides were incubated with blocking buffer for 1 hour at room temperature. Primary antiserum was diluted with blocking buffer to the appropriate working concentration. Blocking buffer was removed and slides were incubated for 18-24 hours at 4oC with primary antiserum. Slides were rinsed 3 times and then incubated with secondary antibodies for 1 hour at room temperature. Slides were again rinsed 3 times and coverslipped. Staining was examined using fluorescence microscopy. Immunocytochemistry: VR1 transfected cells were processed for indirect immunofluorescence. Media was removed and cells were gently washed 3 times with serum-free media. Following fixation procedure, cells were processed for indirect immunofluorescence as described above. Western Blotting: for more information see Guo, A., Vulchanova, L., Wang, J., Li, X., an Elde, R. (1999). Immunocytochemical localization of the vanilloid receptor 1 (VR1): relationship to neuropeptides, the P2X3 purinoceptor and IB4 binding sites. Eur J Neurosci 11, 946-58. Note: Sodium azide (NaN3) interferes with peroxidase reactions and should not be used with peroxidase methodologies. If sodium azide is present in any steps of the staining procedure, the tissue should thoroughly be rinsed with sodium azide-free buffer before performing the peroxidase reaction.
Type: Primary
Antigen: TRPV1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Antigen: TRPV1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
New Product
Limited Time Sale
Select items below are currently on sale
Enter Certificate Details
Inquire for Price
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Item(s) Added to Cart