Anti-XPR1 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX13290
89359-622EA 624.35 CAD
89359-622
Anti-XPR1 Rabbit Polyclonal Antibody
Antibodies
This antibody detects a cell-surface receptor for xenotropic and polytropic murine leukemia viruses that has a possible role in G protein-coupled signal transduction.

IHC-P: Use at a dilution of 1/100 - 1/200. Not tested in other applications.Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: XPR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR