Anti-CD68 Rat Monoclonal Antibody [clone: FA-11]

Supplier: GeneTex
GTX41864
89363-476EA 385.2 CAD
89363-476
Anti-CD68 Rat Monoclonal Antibody [clone: FA-11]
Antibodies
Rat monoclonal antibody [FA-11] to CD68

Recommended Starting Dilutions: Flow Cytometry: 1/50 - 1/100 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD68
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR