Anti-CD7 Mouse Monoclonal Antibody [clone: LT7]

Supplier: GeneTex

GTX74734
89365-546EA 579.45 CAD
89365-546
Anti-CD7 Mouse Monoclonal Antibody [clone: LT7]
Antibodies
Mouse monoclonal antibody [LT7] to CD7

Recommended Starting Dilutions: Flow Cytometry: 1/50 - 1/100 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD7
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR