Anti-CD8 Mouse Monoclonal Antibody [clone: 4B11]

Supplier: GeneTex
GTX75393
89365-788EA 706.94 CAD
89365-788
Anti-CD8 Mouse Monoclonal Antibody [clone: 4B11]
Antibodies
Mouse monoclonal antibody [4B11] to CD8

Recommended Starting Dilutions: Immunohistology - Frozen: 1/20 - 1/40 Western Blotting: Max Dilution 1/25 Immunohistology - Paraffin: 1/20 - 1/40 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD8
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR