Anti-CDX2 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX77810
89366-312EA 732.3 CAD
89366-312
Anti-CDX2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal Antibody to CDX2

WB: Suggested dilution at 0.5-5.0 ug/ml in 5% skim milk / PBS buffer. Optimal dilutions should be determined by end-user.

Type: Primary
Antigen: CDX2
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR