Anti-PGR Mouse Monoclonal Antibody [clone: 4D8]

Supplier: GeneTex
GTX70227
89364-788EA 1052.82 CAD
89364-788
Anti-PGR Mouse Monoclonal Antibody [clone: 4D8]
Antibodies
Mouse monoclonal antibody to human Progesterone receptor

Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: PGR
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR