Anti-PTGDR Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX13088
89359-582EA 677.37 CAD
89359-582
Anti-PTGDR Rabbit Polyclonal Antibody
Antibodies
The prostaglandin D2 receptor is a member of the G-protein-coupled receptor (GPCR) family (subfamily: prostanoid). It has been shown to function as a prostanoid DP receptor. Its activity is mainly mediated by G-S proteins that stimulate adenylate cyclase resulting in an elevation of intracellular cAMP and Ca2+. Knockout studies in mice suggest that the ligand of this receptor, prostaglandin D2 (PGD2), functions as a mast cell-derived mediator to trigger asthmatic responses. Prostaglandin D2 receptor has been reported to be expressed in human small intestine, colon (goblet cells), eye, and eosinophils. ESTs have been isolated from human brain libraries.

IHC-P: Use at a dilution of 1/100 - 1/200. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PTGDR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR