Anti-TNFRSF11A Goat Polyclonal Antibody

GTX10498
89354-056EA 600.34 CAD
89354-056
Anti-TNFRSF11A Goat Polyclonal Antibody
RANK, receptor activator of NF-kappaB, also known as TRANCE receptor and osteoclast differentiation factor (ODFR), is a member of the tumor necrosis factor receptor (TNFR) family. The full-length mouse RANK cDNA encodes a type I transmembrane protein with an extracellular and a cytoplasmic domain. The extracellular domain contains two potential N-linked glycosylation sites. RANK shares significant amino acid homology with other members of the TNFR family in its extracellular four cysteine-rich repeats. Human and murine RANK share 81% amino acid identity in their extracellular domains. RANK is widely expressed, with highest levels in skeletal muscle, thymus, liver, colon, small intestine and adrenal gland. RANK is also expressed in dendritic cells. RANK (receptor activator of NF-kappaB) and RANK ligand (RANKL) are important regulators of interactions between T cells and dendritic cells. RANK is the essential signaling receptor for osteoclast differentiation factor in osteoclastogenesis. Multiple tumor necrosis factor receptor-associated factors (TRAFs) are involved in the signaling of RANK. TRANCE (TNF-related activation-induced cytokines, also known as RANK ligand, osteoprotegerin ligand and osteoclast differentiation factor) is the ligand for RANK. The biological functions mediated by RANK include activation of NF-kappaB and c-jun N-terminal kinase, enhancement of T cell growth and dendritic cell function, induction of osteoclastogenesis and lymph node organogenesis. The soluble form of RANK is able to block TRANCE induced biological activity. The binding of anti-RANK to cell surface RANK triggers signal transduction and induces RANK mediated bioactivity.

Type: Primary
Antigen: TNFRSF11A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR