Anti-S TAG Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX77417
89353-696EA 1058.93 CAD
89353-696
Anti-S TAG Rabbit Polyclonal Antibody
Antibodies
S tag is an epitope tag composed of a 15 residue peptide, KETAAAKFERQHMDS, derived from the pancreatic ribonuclease A.

Type: Primary
Antigen: S TAG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR