Anti-BEX2 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX108673
89355-658EA 623.96 CAD
89355-658
Anti-BEX2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to BEX1/2

Antibody reactive against peptide, detects immunogen. Further validation in progress.

Type: Primary
Antigen: BEX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR