Anti-BID Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX101323
89348-976EA 623.77 CAD
89348-976
Anti-BID Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Bid (BH3 interacting domain death agonist)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IP: 1:100-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: BID
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR