Anti-BCL2L11 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX27888
89362-680EA 749.03 CAD
89362-680
Anti-BCL2L11 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Bim

For WB: Use at a dilution of 1:900 - 1:1,100 dilution. Not tested in other applications. Optimal concentrations/dilutions should be determined by the researcher.

Type: Primary
Antigen: BCL2L11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR