Anti-BCL2L11 Rabbit Monoclonal Antibody

GTX61013
89363-764EA 585.93 CAD
89363-764
Anti-BCL2L11 Rabbit Monoclonal Antibody
Bim, Bcl-2 interacting mediator of cell death, is a pro-apoptotic protein belonging to the BH3-only group of Bcl-2 family members . Bim promotes apoptosis by binding to and inhibiting the activity of anti-apoptotic Bcl-2 family members, inducing the release of cytochrome c from mitochondria . Bim is sequestered in an inactive conformation through binding to the microtubule-associated dynein motor complex. Certain apoptotic stimuli release Bim from microtubules, allowing inhibitory binding to anti-apoptotic Bcl-2 family members and subsequent iniation of apoptosis .

Type: Primary
Antigen: BCL2L11
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR