Anti-BLM Rabbit Polyclonal Antibody

GTX77630
89366-214EA 642.49 CAD
89366-214
Anti-BLM Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to BLM

Recommended Starting Dilutions: Immunoprecipitation: 1ug/mg protein - 4ug/mg protein Western Blotting: 1/1,000 - 1/10,000 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: BLM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR