Anti-BLOOD GROUP A Mouse Monoclonal Antibody [clone: HE-10]

Supplier: GeneTex

GTX22523
89361-266EA 565.64 CAD
89361-266
Anti-BLOOD GROUP A Mouse Monoclonal Antibody [clone: HE-10]
Antibodies
Type: Primary
Antigen: BLOOD GROUP A
Clonality: Monoclonal
Clone: [HE-10]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR