Anti-BMP2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX103626
89350-050EA 264.05 CAD
89350-050
Anti-BMP2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to BMP2 (bone morphogenetic protein 2)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: BMP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR